missing translation for 'onlineSavingsMsg'
Learn More

Glutaraldehyde, 50% Aqueous Solution, MP Biomedicals

Product Code. 11442731 Shop All MP Biomedicals Products
Change view
Click to view available options
Quantity:
100 mL
Unit Size:
100mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11442731 100 mL 100mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 11442731 Supplier MP Biomedicals Supplier No. 0219859580
 Restricted Item
Estimated Shipment: 04-02-2027
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Specifications

Physical Form Aqueous Solution
CAS 111-30-8
Density 1.061 g/cm3 (at 20 °C)
Quantity 100 mL
Molecular Formula C5H8O2
MDL Number MFCD00007025
Synonym glutaraldehyde, glutaral, glutaric dialdehyde, cidex, glutardialdehyde, 1,5-pentanedial, sonacide, glutarol, aldesan, glutaric aldehyde
InChI Key SXRSQZLOMIGNAQ-UHFFFAOYSA-N
SMILES O=CCCCC=O
IUPAC Name pentanedial
Molecular Weight (g/mol) 100.12
PubChem CID 3485
ChEBI CHEBI:64276
Formula Weight 100.11
Concentration or Composition (by Analyte or Components) 50%
Chemical Name or Material Glutaraldehyde
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.