missing translation for 'onlineSavingsMsg'
Learn More

Glutaric acid, 99%

Product Code. 10436730 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
25 g
100 g
500 g
Packaging:
Glass Bottle
Plastic bottle
Unit Size:
100g
25g
500g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
10436730 25 g Glass Bottle 25g
10122820 100 g Plastic bottle 100g
10081060 500 g Plastic bottle 500g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 10436730 Supplier Thermo Scientific Acros Supplier No. 119960250
 Restricted Item
Estimated Shipment: 22-10-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 110-94-1 | C5H8O4 | 132.12 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 110-94-1
Molecular Formula C5H8O4
Molecular Weight (g/mol) 132.12
MDL Number MFCD00004410
InChI Key JFCQEDHGNNZCLN-UHFFFAOYSA-N
Synonym glutaric acid, 1,5-pentanedioic acid, 1,3-propanedicarboxylic acid, pentandioic acid, glutarate, n-pyrotartaric acid, unii-h849f7n00b, propane-1,3-dicarboxylic acid, c4-c6 dibasic acids, carboxylic acids, c6-18 and c5-15-di
PubChem CID 743
ChEBI CHEBI:17859
IUPAC Name pentanedioic acid
SMILES OC(=O)CCCC(O)=O

Specifications

Melting Point 92.0°C to 99.0°C
Color Orange
Boiling Point 302.0°C
Infrared Spectrum Authentic
Assay Percent Range 99%
Packaging Glass Bottle
Linear Formula HO2C(CH2)3CO2H
Quantity 25 g
Beilstein 02, 631
Fieser 01,412
Merck Index 15, 4509
Solubility Information Solubility in water: 430g/L (20°C). Other solubilities: freely soluble in abs. alcohol and ether, soluble in benzene and chloroform
Formula Weight 132.12
Percent Purity 99%
Physical Form Crystalline Powder
Chemical Name or Material Glutaric acid, 99%
Show More Show Less
compliance-icons
Product Identifier
  • Glutaric acid
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
Hazard Statement
  • H319-Causes serious eye irritation.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P311-Call a POISON CENTER/doctor/ .
Supplemental information
  • MIXTURE LIST-Contain: Glutaric acid

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.