missing translation for 'onlineSavingsMsg'
Learn More

Glutaric dialdehyde, 50 wt.% solution in water

Product Code. 10533064 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
1 kg
2.5 kg
10 kg
Packaging:
Glass bottle
Plastic drum
Unit Size:
10kg
1kg
2.5kg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
10533064 1 kg Glass bottle 1kg
10628283 2.5 kg Glass bottle 2.5kg
10454425 10 kg Plastic drum 10kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 10533064 Supplier Thermo Scientific Acros Supplier No. 410960010
  • ONLINE EXCLUSIVE PRICE
    85.95 €
    80.20 € / 1kg
    Save 5.75 €
    6% Off
View all promotions
 Restricted Item
In Stock
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 111-30-8 | C5H8O2 | 100.12 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Glutaric dialdehyde
Name Note 50 wt% Solution in Water
CAS 111-30-8, 7732-18-5
Melting Point -33.0°C
Density 1.1300g/mL
pH 3.2 to 4.2
Boiling Point 101.5°C (740.0 mmHg)
Packaging Glass bottle
Molecular Formula C5H8O2
Linear Formula OHC(CH2)3CHO
MDL Number MFCD00007025
Quantity 1 kg
Beilstein 01, 776
Fieser 01,411
Merck Index 15, 4508
Specific Gravity 1.13
Synonym glutaraldehyde, glutaral, glutaric dialdehyde, cidex, glutardialdehyde, 1,5-pentanedial, sonacide, glutarol, aldesan, glutaric aldehyde
Solubility Information Solubility in water: soluble
InChI Key SXRSQZLOMIGNAQ-UHFFFAOYSA-N
SMILES O=CCCCC=O
IUPAC Name pentanedial
Molecular Weight (g/mol) 100.12
PubChem CID 3485
ChEBI CHEBI:64276
Viscosity 20 mPa.s (50°C)
Formula Weight 100.12
Physical Form Solution
Show More Show Less
compliance-icons compliance-icons compliance-icons compliance-icons
Product Identifier
  • Glutaric dialdehyde, 50 wt.% solution in water
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 3
  • Skin corrosion/irritation Category 1 B
  • Serious eye damage/eye irritation Category 1
  • Respiratory sensitiser Category 1
  • Skin sensitiser Category 1
  • ACUTE AQUATIC Acute 1
  • LONG-TERM AQUATIC HAZARD Chronic 2
Hazard Statement
  • H301-Toxic if swallowed.
  • H331-Toxic if inhaled.
  • H314-Causes severe skin burns and eye damage.
  • H317-May cause an allergic skin reaction.
  • H334-May cause allergy or asthma symptoms or breathing difficulties if inhaled.
  • H400-Very toxic to aquatic life.
  • H411-Toxic to aquatic life with long lasting effects.
  • EUH071-Corrosive to the respiratory tract.
Precautionary Statement
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.