missing translation for 'onlineSavingsMsg'
Learn More

Glutathione (glycine-13C2,15N) Trifluoroacetate Salt, TRC

Product Code. 30006090
Change view
Click to view available options
Quantity:
1 mg
2.5 mg
Unit Size:
1mg
2.5mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30006090 2.5 mg 2.5mg
30010279 1 mg 1mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30006090 Supplier Toronto Research Chemicals Supplier No. TRCG5979542.5MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

Molecular Formula C813C2H17N215NO6S • C2HF3O2
Molecular Weight (g/mol) 310.3011402
Synonym L-Glutamyl-L-cysteinyl-glycine-3C2,15N Trifluoroacetate Salt
SMILES O[13C]([13C]([H])([15N](C([C@@H](NC(CC[C@H](N)C(O)=O)=O)CS)=O)[H])[H])=O.FC(F)(C(O)=O)F

Specifications

Quantity 2.5 mg
InChI Formula InChI=1S/C10H17N3O6S.C2HF3O2/c11-5(10(18)19)1-2-7(14)13-6(4-20)9(17)12-3-8(15)16;3-2(4,5)1(6)7/h5-6,20H,1-4,11H2,(H,12,17)(H,13,14)(H,15,16)(H,18,19);(H,6,7)/t5-,6-;/m0./s1/i3+1,8+1,12+1;
Percent Purity >95%
Chemical Name or Material Glutathione (glycine-13C2,15N) Trifluoroacetate Salt
Grade HPLC
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.