Learn More
Glutathione Peroxidase 1/GPX1 Goat anti-Human, Mouse, Rat, Polyclonal, R&D Systems™
GREENER_CHOICE
Goat Polyclonal Antibody has been used in 15 publications
Brand:  R&D Systems AF3798-SP
Encompass
Product Code. 15742234
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products »
Description
Description
Glutathione Peroxidase 1/GPX1 Polyclonal specifically detects Glutathione Peroxidase 1/GPX1 in Human, Mouse, Rat samples. It is validated for Western Blot, Simple Western, Immunohistochemistry, Knockout Validated.