missing translation for 'onlineSavingsMsg'
Learn More

Glutathione Reduced, Cell Culture Reagent, ≥98%, MP Biomedicals™

Product Code. 11465202 Shop All MP Biomedicals Products
Change view
Click to view available options
Quantity:
100 g
Unit Size:
100g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11465202 100 g 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 11465202 Supplier MP Biomedicals Supplier No. 0219467980
  • ONLINE EXCLUSIVE PRICE
    1135.00 €
    1073.10 € / 100g
    Save 61.90 €
    5% Off
 Restricted Item
Estimated Shipment: 23-12-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Glutathione Reduced, Cell Culture Reagent, ≥98% is used for protein purification and elutes GST-tagged protein from glutathione agarose.

  • Glutathione suppresses human immunodeficiency virus expression in chronically infected monocytic cells.
  • It is a useful tripeptide involved in many aspects of metabolism, including transport of g-glutanyl amino acids and reductive cleavage of disulfide bonds.
  • Glutathione is an endogenous antioxidant that plays a major role in reducing reactive oxygen species formed during cellular metabolism and the respiratory burst.
  • Glutathione-S-transferase catalyzes the formation of glutathione thioethers with xenobiotics, leukotrienes, and other molecules that have an electrophilic center.
  • Glutathione also forms disulfide bonds with cysteine residues in proteins.
  • Via these mechanisms, it can have the paradoxical effect of reducing the efficacy of anti-cancer agents.
  • Typical Working Concentration: May be used at 5-10 mM to elute glutathione S-transferase (GST) from glutathione agarose.

Chemical Identifiers

CAS 70-18-8
Molecular Formula C10H17N3O6S
Molecular Weight (g/mol) 307.321
MDL Number MFCD00065939
InChI Key RWSXRVCMGQZWBV-WDSKDSINSA-N
Synonym glutathione, l-glutathione, glutathion, glutathione-sh, glutinal, isethion, tathion, reduced glutathione, deltathione, neuthion
PubChem CID 124886
ChEBI CHEBI:16856
IUPAC Name (2S)-2-amino-5-[[(2R)-1-(carboxymethylamino)-1-oxo-3-sulfanylpropan-2-yl]amino]-5-oxopentanoic acid
SMILES C(CC(=O)NC(CS)C(=O)NCC(=O)O)C(C(=O)O)N

Specifications

Melting Point 195°C
Color White
Assay Percent Range >98%
Quantity 100 g
Solubility Information 292.5 mg/mL
Formula Weight 307.32
Percent Purity ≥98%
Grade Cell Culture
Physical Form Powder
Chemical Name or Material Glutathione Reduced, Cell Culture Reagent

Unless specified otherwise, MP Biomedical's products are for research or further manufacturing use only, not for direct human use.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.