missing translation for 'onlineSavingsMsg'
Learn More

Glutathione Reduced, Fisher BioReagents

Product Code. 11483074
Change view
Click to view available options
Quantity:
1 g
5 g
10 g
25 g
50 g
100 g
Packaging:
Amber Glass
Unit Size:
100g
10g
1g
25g
50g
5g
6 product options available for selection
Product selection table with 6 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
11483074 50 g Amber Glass 50g
10071213 1 g Amber Glass 1g
10562785 5 g Amber Glass 5g
10204873 10 g Amber Glass 10g
10482475 25 g Amber Glass 25g
10566394 100 g Amber Glass 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 6 options available.
6 options
This item is not returnable. View return policy
Product Code. 11483074 Supplier Fisher BioReagents Supplier No. BP252150
Request Bulk or Custom Format

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy
Glutathione is a useful tripeptide involved in many aspects of metabolism, including transport of γ-glutamyl amino acids and reductive cleavage of disulfide bonds.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 70-18-8
Molecular Formula C10H17N3O6S
Molecular Weight (g/mol) 307.321
InChI Key RWSXRVCMGQZWBV-WDSKDSINSA-N
Synonym glutathione, l-glutathione, glutathion, glutathione-sh, glutinal, isethion, tathion, reduced glutathione, deltathione, neuthion
PubChem CID 124886
ChEBI CHEBI:16856
IUPAC Name (2S)-2-amino-5-[[(2R)-1-(carboxymethylamino)-1-oxo-3-sulfanylpropan-2-yl]amino]-5-oxopentanoic acid
SMILES C(CC(=O)NC(CS)C(=O)NCC(=O)O)C(C(=O)O)N

Specifications

Melting Point 182°C
Color White
pH 3
Ignition Residue 0.01% max.
Loss on Drying Report %
Assay Percent Range ≥98 %
Packaging Amber Glass
Quantity 50 g
Merck Index 15, 4511
Specific Rotation -21 to -11° (+ or -)
Formula Weight 307.3
Percent Purity ≥98%
Physical Form Powder or Solid
Chemical Name or Material Glutathione Reduced
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.