missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Glutathione Reductase Fluorescent Activity Kit

Product Code. 15830903
Click to view available options
Quantity:
96 tests
Unit Size:
Pack of 96
This item is not returnable. View return policy

Product Code. 15830903

Brand: Invitrogen™ EIAGRF

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Includes

96 Well Plates, standard, detection reagent, assay buffer, substrate, and lot-specific technical data sheet.

Assay Glutathione Reductase Activity in Serum, Plasma, RBCs and Cell Lysates.

  • Assay Type: Enzyme Activity kit
  • Sample Types: Serum, Plasma, RBCs and Cell Lysates
  • Sensitivity: 0.009 mU/mL
  • Standard Curve Range: 0.15 mU/mL–5 mU/mL
TRUSTED_SUSTAINABILITY

Specifications

Content And Storage 96-well plate(s), standard, detection reagent, assay buffer, substrate, and lot-specific technical data sheet.

Store at –20°C.
Immunoassay Kit Format Assay Activity Kit
Product Type Glutathione Reductase Fluorescent Activity Kit
Sample Type Cell Culture Supernatant, Plasma, Serum
For Use With (Equipment) Microplate Reader
Quantity 96 tests
Shipping Condition Approved for shipment on Wet or Dry Ice
Target Kit Named Glutathione Reductase

For Research Use Only. Not for use in diagnostic procedures.

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.