missing translation for 'onlineSavingsMsg'
Learn More

Gly-Gly, JustPURE™, white powder, Fisher BioReagents

Product Code. 10838830
Change view
Click to view available options
Quantity:
50 g
Unit Size:
50g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
10838830 50 g 50g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 10838830 Supplier Fisher BioReagents Supplier No. BP293050
Request Bulk or Custom Format

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Specifications

CAS 556-50-3
Color White
pH 5.5 to 6.0
Physical Form Powder
Quantity 50 g
Packaging Poly Bottle
Synonym Diglycine
Solubility Information Soluble in Water
Molecular Weight (g/mol) 132.119
Grade JustPURE
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.