missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals Gly-Phe beta-naphthylamide

Product Code. 15446539
Change view
Click to view available options
Quantity:
100 mg
Unit Size:
100mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15446539 100 mg 100mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 15446539 Supplier Thermo Scientific Chemicals Supplier No. J64718.MC
 Restricted Item
In Stock
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

347.41 g/mol

Gly-Phe beta-naphthylamide is used to accumulate inside lysosomes leading to their destruction. Studies report that Gly-Phe-β-naphthylamide significantly inhibits the activation of caspase-8, and has also been used to study recombinant cathepsin C.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Specifications

Purification Method HPLC
Common Name Glycylphenylalanine 2-naphthylamide
Content And Storage -30°C to -10°C
Format Crystalline
For Use With (Application) Substrate for cathepsin C
Molecular Weight (g/mol) 347.41
Product Type Gly-Phe beta-naphthylamide
Quantity 100 mg
Regulatory Status RUO
Sequence (S)-2-(2-Aminoacetamido)-N-(naphthalen-2-yl)-3-phenylpropanamide

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.