missing translation for 'onlineSavingsMsg'
Learn More

Glycerine, BAKER ANALYZED™, J.T. Baker™

Product Code. 10269620 Shop All J.T.Baker Products
Change view
Click to view available options
Quantity:
1 L
Unit Size:
1 L
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
10269620 1 L 1 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 10269620 Supplier JT Baker Supplier No. 7044.1000
  • ONLINE EXCLUSIVE PRICE
    179.70 €
    158.99 € / 1 L
    Save 20.71 €
    11% Off
 Restricted Item
Estimated Shipment: 09-12-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Laboratory Reagent

Specifications

Melting Point 18.6°C
CAS 56-81-5
Color Passes Test
Density 1.26 g/cm³ (20°C)
Quantity 1 L
Flash Point 177°C
Packaging Plastic Bottle
Assay Percent Range min. 99.5%
Molecular Formula C3H8O3
MDL Number MFCD00004722
Synonym Glycerol , 1,2,3-Propanetriol
Boiling Point 290°C (1013 hPa)
InChI Key PEDCQBHIVMGVHV-UHFFFAOYSA-N
SMILES OCC(O)CO
IUPAC Name propane-1,2,3-triol
Molecular Weight (g/mol) 92.09
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.