missing translation for 'onlineSavingsMsg'
Learn More

Glycerine Trioleate (Technical Grade), TRC

Product Code. 30389785
Change view
Click to view available options
Quantity:
25 g
Unit Size:
25g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30389785 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30389785 Supplier Toronto Research Chemicals Supplier No. TRCG59822525G
 Restricted Item
Estimated Shipment: 18-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Triolein
CAS 122-32-7
Molecular Formula C57 H104 O6
Quantity 25 g
Synonym 9-Octadecenoic acid (9Z)-, 1,1',1''-(1,2,3-propanetriyl) ester, 9-Octadecenoic acid (9Z)-, 1,2,3-propanetriyl ester (9CI), 9-Octadecenoic acid (Z)-, 1,2,3-propanetriyl ester, Olein, tri- (8CI), Actor LO 1, Aldo TO, Edenor NHTi-G, Emerest 2423, Emery 2423, Emery oleic acid ester 2230, Estol 1433, Glycerin trioleate, Glycerol trioleate, Glycerol triolein, Glyceryl trioleate, Glyceryl-1,2,3-trioleate, Kaolube 190, Kemester 1000, OOO triacylglycerol, Oleic acid triglyceride, Oleic triglyceride, Oleyl triglyceride, Radia 7363, Raoline, Triglyceride OOO, Triolein, Trioleoylglyceride, Trioleoylglycerol, sn-Glyceryl trioleate
InChI Formula InChI=1S/C57H104O6/c1-4-7-10-13-16-19-22-25-28-31-34-37-40-43-46-49-55(58)61-52-54(63-57(60)51-48-45-42-39-36-33-30-27-24-21-18-15-12-9-6-3)53-62-56(59)50-47-44-41-38-35-32-29-26-23-20-17-14-11-8-5-2/h25-30,54H,4-24,31-53H2,1-3H3/b28-25-,29-26-,30-27-
SMILES CCCCCCCC\C=C/CCCCCCCC(=O)OCC(COC(=O)CCCCCCC\C=C/CCCCCCCC)OC(=O)CCCCCCC\C=C/CCCCCCCC
IUPAC Name 2,3-bis[[(Z)-octadec-9-enoyl]oxy]propyl (Z)-octadec-9-enoate
Molecular Weight (g/mol) 885.43
Formula Weight 884.7833
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.