missing translation for 'onlineSavingsMsg'
Learn More

Glycerol, 99.5%, for molecular biology, DNAse, RNAse and Protease free

Product Code. 10692372 Shop All Thermo Scientific Products
Change view
Click to view available options
Quantity:
500 mL
Unit Size:
500mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
10692372 500 mL 500mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 10692372 Supplier Thermo Scientific Supplier No. 327255000
  • ONLINE EXCLUSIVE PRICE
    108.00 €
    104.00 € / 500mL
    Save 4.00 €
    3% Off
 Restricted Item
In Stock
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 56-81-5 | C3H8O3 | 92.09 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Specifications

CAS 56-81-5
Absorbance (0.5M in water), (0.5 M in water) at 0nm, 0.02 max. at 280nm, 0.06 max. at 260nm
Melting Point 18.0°C
Density 1.2610g/mL
pH 6 to 7 (10% soln. at 25°C)
Boiling Point 290.0°C
Flash Point 160°C
Assay Percent Range 99.5% min. (GC)
Packaging Glass Bottle
Molecular Formula C3H8O3
Linear Formula HOCH2CH(OH)CH2OH
Refractive Index 1.4716
MDL Number MFCD00004722
Quantity 500 mL
Beilstein 01, 502
Merck Index 15, 4520
Specific Gravity 1.261
Synonym glycerol, glycerin, glycerine, 1,2,3-propanetriol, glycyl alcohol, trihydroxypropane, glyceritol, propanetriol, 1,2,3-trihydroxypropane, osmoglyn
Solubility Information Solubility in water: >500g/L (20°C). Other solubilities: soluble in methanol, ethanol and 2-propanol, insoluble in ether, benzene, chloroform
InChI Key PEDCQBHIVMGVHV-UHFFFAOYSA-N
SMILES OCC(O)CO
IUPAC Name propane-1,2,3-triol
Molecular Weight (g/mol) 92.09
PubChem CID 753
ChEBI CHEBI:17754
Viscosity 1069 mPa.s (20°C)
Formula Weight 92.09
Percent Purity 99.5%
Grade Molecular Biology
Physical Form Liquid
Chemical Name or Material Glycerol
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.