missing translation for 'onlineSavingsMsg'
Learn More

Glycerol, >99%, Highly Purified, MP Biomedicals™

Product Code. 11485202 Shop All MP Biomedicals Products
Change view
Click to view available options
Quantity:
100 mL
500 mL
1 L
Unit Size:
1 L
100mL
500mL
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11485202 500 mL 500mL
11401112 100 mL 100mL
11421112 1 L 1 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11485202 Supplier MP Biomedicals Supplier No. 0219468090
  • ONLINE EXCLUSIVE PRICE
    178.00 €
    168.00 € / 500mL
    Save 10.00 €
    5% Off
 Restricted Item
In Stock
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 56-81-5 | C3H8O3 | 92.09 g/mol

Chemical Identifiers

CAS 56-81-5
Molecular Formula C3H8O3
Molecular Weight (g/mol) 92.09
MDL Number MFCD00004722
InChI Key PEDCQBHIVMGVHV-UHFFFAOYSA-N
Synonym glycerol, glycerin, glycerine, 1,2,3-propanetriol, glycyl alcohol, trihydroxypropane, glyceritol, propanetriol, 1,2,3-trihydroxypropane, osmoglyn
PubChem CID 753
ChEBI CHEBI:17754
IUPAC Name propane-1,2,3-triol
SMILES OCC(O)CO

Specifications

Color Colorless
Density 1.2613 g/cu cm at 20°C (Lit.)
Assay Percent Range >99%
Odor Odorless
Quantity 500 mL
Formula Weight 92.09
Grade Not Available
Physical Form Syrupy Liquid
Chemical Name or Material Glycerol
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.