missing translation for 'onlineSavingsMsg'
Learn More

Glycerol, for analysis, 86-88% wt% aqueous solution

Product Code. 10441301 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
250 mL
1 L
2.5 L
10 L
Packaging:
Glass bottle
Plastic drum
Unit Size:
1 L
10 L
2.5 L
250mL
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10441301 1 L Glass bottle 1 L
10451301 250 mL Glass bottle 250mL
10665521 2.5 L Glass bottle 2.5 L
10404035 10 L Plastic drum 10 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
This item is not returnable. View return policy
Product Code. 10441301 Supplier Thermo Scientific Acros Supplier No. 295600010
  • ONLINE EXCLUSIVE PRICE
    106.00 €
    102.00 € / 1 L
    Save 4.00 €
    3% Off
 Restricted Item
Estimated Shipment: 25-08-2026
Add to basket
This item is not returnable. View return policy

CAS: 56-81-5 | C3H8O3 | 92.09 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 56-81-5 , 7732-18-5
Molecular Formula C3H8O3
Molecular Weight (g/mol) 92.09
MDL Number MFCD00004722
InChI Key PEDCQBHIVMGVHV-UHFFFAOYSA-N
Synonym glycerol, glycerin, glycerine, 1,2,3-propanetriol, glycyl alcohol, trihydroxypropane, glyceritol, propanetriol, 1,2,3-trihydroxypropane, osmoglyn
PubChem CID 753
ChEBI CHEBI:17754
IUPAC Name propane-1,2,3-triol
SMILES OCC(O)CO

Specifications

Melting Point 18.0°C
Color Colorless
Density 1.2270g/mL
Boiling Point 290.0°C
Flash Point 160°C
Infrared Spectrum Authentic
Packaging Glass bottle
Linear Formula HOCH2CH(OH)CH2OH
Refractive Index 1.4520 to 1.4750
Quantity 1 L
Beilstein 01, 502
Merck Index 15, 4520
Specific Gravity 1.227
Solubility Information Solubility in water: >500g/L (20°C). Other solubilities: soluble in methanol, ethanol and 2-propanol, insoluble in ether, benzene, chloroform and in, fixed and volatile oils
Viscosity 1069 mPa.s (20°C)
Formula Weight 92.09
Percent Purity 86.0 to 88.0%
Grade Analysis
Physical Form Viscous Liquid
Chemical Name or Material Glycerol, 86 to 88% wt% Aqueous Solution
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.