missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Glycerol, Anhydrous, Ultrapure Bioreagent, J.T. Baker™
Click to view available options
Quantity:
500 mL
4 L
Packaging:
HDPE Pressure Bottom Poly Bottle
Poly Bottle
Description
- For enzyme stabilization, freezing and electrophoresis buffers
Specifications
Flash Point | 199.00°C |
Packaging | HDPE Pressure Bottom Poly Bottle |
Quantity | 500 mL |
Specific Gravity | 1L = 1.26kg |
Formula Weight | 92.1 |
Grade | Ultrapure Bioreagent |
Chemical Name or Material | Glycerol, Anhydrous |
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'