missing translation for 'onlineSavingsMsg'
Learn More

Glycerol, Anhydrous, Ultrapure Bioreagent, J.T. Baker™

Product Code. 15558224 Shop All J.T.Baker Products
Change view
Click to view available options
Quantity:
500 mL
4 L
Packaging:
HDPE Pressure Bottom Poly Bottle
Poly Bottle
Unit Size:
Each
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
15558224 4 L Poly Bottle Each
15548224 500 mL HDPE Pressure Bottom Poly Bottle Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 15558224 Supplier Avantor J.T.Baker Supplier No. 404301

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High purity reagents tested for use in biotechnology applications, such as electrophoresis, and liquid chromatography

    For enzyme stabilization, freezing and electrophoresis buffers; For Laboratory,Research,or Manufacturing Use

Chemical Identifiers

CAS 56-81-5
Molecular Formula C3H8O3
Molecular Weight (g/mol) 92.09
MDL Number MFCD00004722
InChI Key PEDCQBHIVMGVHV-UHFFFAOYSA-N
Synonym glycerol, glycerin, glycerine, 1,2,3-propanetriol, glycyl alcohol, trihydroxypropane, glyceritol, propanetriol, 1,2,3-trihydroxypropane, osmoglyn
PubChem CID 753
ChEBI CHEBI:17754
IUPAC Name propane-1,2,3-triol
SMILES OCC(O)CO

Specifications

Flash Point 199.00°C
Packaging Poly Bottle
Quantity 4 L
Specific Gravity 1L = 1.26kg
Formula Weight 92.1
Grade Ultrapure Bioreagent
Chemical Name or Material Glycerol, Anhydrous
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.