missing translation for 'onlineSavingsMsg'
Learn More

Glycerol formal, 99+%, mixture of isomers, stabilized

Product Code. 10063270 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
5 mL
250 mL
500 mL
Packaging:
Glass bottle
Unit Size:
250mL
500mL
5mL
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
10063270 500 mL Glass bottle 500mL
10083230 5 mL Glass bottle 5mL
10490511 250 mL Glass bottle 250mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 10063270 Supplier Thermo Scientific Acros Supplier No. 259775000
ChemsChrom Mega Deals
Use promo code 27993
View all promotions
 Restricted Item
Estimated Shipment: 07-12-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 4740-78-7 | C4H8O3 | 104.11 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Glycerol formal
Name Note 99+%
CAS 4740-78-7, 5464-28-8
Density 1.2100g/mL
pH 4.0 to 6.5 (10% soln.)
Boiling Point 193.0°C to 195.0°C
Flash Point 97°C
Infrared Spectrum Authentic
Assay Percent Range 99% min. Sum of alpha-and beta-isomers (GC)
Packaging Glass bottle
Molecular Formula C4H8O3
Refractive Index 1.4500 to 1.4520
MDL Number MFCD00003218
Quantity 500 mL
Merck Index 14, 4485
Specific Gravity 1.21
Solubility Information (50% in water) Clear
Molecular Weight (g/mol) 104.11
Viscosity 16 mPa.s (20°C)
Formula Weight 104.11
Percent Purity 99+%
Physical Form Liquid
Show More Show Less
compliance-icons compliance-icons
Product Identifier
  • Glycerol formal, mixture of approx. 55% 5-Hydroxy-1,3-dioxane and approx. 45% 4-Hydroxymethyl-1,3-dioxolane
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Reproductive toxicity Category 2
Hazard Statement
  • H361-Suspected of damaging fertility or the unborn child.
  • H319-Causes serious eye irritation.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P308+P313-IF exposed or concerned: Get medical advice/attention.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.