missing translation for 'onlineSavingsMsg'
Learn More

Glycerol, Molecular Biology Grade

Product Code. 15473719
Change view
Click to view available options
Quantity:
500 mL
1 L
Unit Size:
1 L
500mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15473719 1 L 1 L
15463719 500 mL 500mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 15473719 Supplier Thermo Scientific Alfa Aesar Supplier No. J61059.K2
 Restricted Item
In Stock
Add to basket
Edge

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Useful as a cryoprotectant. Glycerol is evolved in the storage and concentration of enzymes. It plays a vital role in sample preparation and gel formation for polyacrylamide gel electrophoresis. It is also used to aid in casting gradient gels, protein stabilizer and storage buffer component. | CAS: 56-81-5 | C3H8O3 | 92.09 g/mol

Useful as a cryoprotectant.Glycerol is evolved in the storage and concentration of enzymes. It plays a vital role in sample preparation and gel formation for polyacrylamide gel electrophoresis. It is also used to aid in casting gradient gels, protein stabilizer and storage buffer component. Further, it is used as a solvent, emollient, pharmaceutical agent and sweetening agent.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 56-81-5
Molecular Formula C3H8O3
Molecular Weight (g/mol) 92.09
MDL Number MFCD00004722
InChI Key PEDCQBHIVMGVHV-UHFFFAOYSA-N
Synonym glycerol, glycerin, glycerine, 1,2,3-propanetriol, glycyl alcohol, trihydroxypropane, glyceritol, propanetriol, 1,2,3-trihydroxypropane, osmoglyn
PubChem CID 753
ChEBI CHEBI:17754
IUPAC Name propane-1,2,3-triol
SMILES OCC(O)CO

Specifications

Melting Point 18°C
Density 1.26
Boiling Point 182°C (20 mmHg)
Flash Point 160°C (320°F)
Packaging Plastic bottle
Linear Formula HOCH2CH(OH)CH2OH
Refractive Index 1.474
Quantity 1 L
Beilstein 635685
Sensitivity Hygroscopic
Specific Gravity 1.26
Solubility Information Miscible with water and alcohol. Partially miscible with ether and ethyl acetate. Immiscible with chloroform.
Formula Weight 92.09
Grade Molecular Biology
Physical Form Liquid
Chemical Name or Material Glycerol
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.