missing translation for 'onlineSavingsMsg'
Learn More

Glycerol monostearate, purified

Product Code. 11324918
Change view
Click to view available options
Quantity:
250 g
1 kg
5 kg
Unit Size:
1kg
250g
5kg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11324918 1 kg 1kg
11304918 250 g 250g
11314918 5 kg 5kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11324918 Supplier Thermo Scientific Alfa Aesar Supplier No. 043883.A1
 Restricted Item
Add to basket
Edge

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 31566-31-1 | C21H42O4 | 358.563 g/mol

Glycerol monostearate is used as an emulsifier, resin lubricant, opacifier, emollient, bodying agent in a variety of cosmetic formulations for skincare and haircare. It is also used as a thickening, anti-caking and preservative agent. It is also useful for preventing ice creams from drying out or being too sweet. It is further used as a foaming agent for the foam-mat drying of papaya. It is also used as an anti-staling agent in bread.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 31566-31-1
Molecular Formula C21H42O4
Molecular Weight (g/mol) 358.563
MDL Number MFCD00036186
InChI Key VBICKXHEKHSIBG-UHFFFAOYSA-N
Synonym glyceryl monostearate, monostearin, glyceryl stearate, 1-stearoyl-rac-glycerol, tegin, 1-monostearin, 1-glyceryl stearate, glycerol monostearate, 1-monostearoylglycerol, octadecanoic acid, 2,3-dihydroxypropyl ester
PubChem CID 24699
ChEBI CHEBI:75555
IUPAC Name 2,3-dihydroxypropyl octadecanoate
SMILES CCCCCCCCCCCCCCCCCC(=O)OCC(CO)O

Specifications

Quantity 1 kg
Merck Index 14,4489
Solubility Information Soluble in hot organic solvents;Soluble in hot water; Slightly soluble in ethanol; Insoluble in aliphatic solvents.
Formula Weight 358.56
Physical Form Powder
Chemical Name or Material Glycerol monostearate, Purified

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.