missing translation for 'onlineSavingsMsg'
Learn More

Glycerol Phosphate Disodium Salt Hydrate, Isomeric Mixture, TRC

Product Code. 30135904
Change view
Click to view available options
Quantity:
1 g
5 g
10 g
Unit Size:
10g
1g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30135904 5 g 5g
30136195 10 g 10g
30011055 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30135904 Supplier Toronto Research Chemicals Supplier No. TRCG6016035G
 Restricted Item
Estimated Shipment: 28-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 55073-41-1
Molecular Formula C3H7Na2O6P • xH2O
Molecular Weight (g/mol) 216.04
Synonym 1,2,3-Propanetriol, mono(dihydrogen phosphate), disodium salt, hydrate (9CI), Sodium glycerophosphate hydrated, Glycerylphosphate sodium hydrate, Glycerol monophosphate disodium salt hydrate, Disodium glycerophosphate hydrate
SMILES OCC(OP(O[Na])(O[Na])=O)CO

Specifications

Quantity 5 g
Formula Weight 449.9657
Chemical Name or Material Glycerol Phosphate Disodium Salt Hydrate, Isomeric Mixture
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.