missing translation for 'onlineSavingsMsg'
Learn More

Glycerol, puriss., Honeywell
BIOPHARMA

Product Code. p-7073755
Click to view available options
Quantity:
1 L
5 L
70 kg
250 kg
Unit Size:
1 L
250kg
5 L
70kg
This item is not returnable. View return policy

Product Code. 15681470

Brand: Honeywell Riedel-de Haën 155231L

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Chemical Identifiers

CAS 56-81-5
Molecular Formula C3H8O3
Molecular Weight (g/mol) 92.09
MDL Number MFCD00004722
InChI Key PEDCQBHIVMGVHV-UHFFFAOYSA-N
Synonym glycerol, glycerin, glycerine, 1,2,3-propanetriol, glycyl alcohol, trihydroxypropane, glyceritol, propanetriol, 1,2,3-trihydroxypropane, osmoglyn
PubChem CID 753
ChEBI CHEBI:17754
IUPAC Name propane-1,2,3-triol
SMILES OCC(O)CO

Specifications

Density 1.2580 to 1.2644g/mL at 20°C, ≥1.257g/mL at 25°C
Flash Point 160°C (320°F)
Ignition Residue ≤0.01% (as SO4)
Ignition Point 698°F
Linear Formula HOCH2CH(OH)CH2OH
Quantity 1 L
UN Number NONH for all modes of transport
Beilstein 635685
Vapor Density 3.1 (vs air)
Formula Weight 92.09g/mol
Vapor Pressure 1mmHg (20°C)
Percent Purity 99 to 101% (alkalimetric)
Grade puriss.
Chemical Name or Material Glycerol, Meets analytical specification of Ph. Eur. BP, USP, FCC.
Show More Show Less
Product Identifier
  • Glycerol
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.