missing translation for 'onlineSavingsMsg'
Learn More

Glycerol, puriss. p.a., ACS reagent, Solstice

Product Code. 15685690
Change view
Click to view available options
Quantity:
250 mL
1 L
2.5 L
Unit Size:
1 L
2.5 L
250mL
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15685690 250 mL 250mL
15665690 1 L 1 L
15675690 2.5 L 2.5 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 15685690 Supplier Fluka Supplier No. 49770250ML
 Restricted Item
Estimated Shipment: 07-12-2026
Add to basket
This item is not returnable. View return policy

Chemical Identifiers

CAS 56-81-5
Molecular Formula C3H8O3
Molecular Weight (g/mol) 92.09
MDL Number MFCD00004722
InChI Key PEDCQBHIVMGVHV-UHFFFAOYSA-N
Synonym glycerol, glycerin, glycerine, 1,2,3-propanetriol, glycyl alcohol, trihydroxypropane, glyceritol, propanetriol, 1,2,3-trihydroxypropane, osmoglyn
PubChem CID 753
ChEBI CHEBI:17754
IUPAC Name propane-1,2,3-triol
SMILES OCC(O)CO

Specifications

Density 1.25g/mL
Flash Point 160°C (320°F)
Ignition Point 698°F
Linear Formula HOCH2CH(OH)CH2OH
Quantity 250 mL
UN Number NONH for all modes of transport
Beilstein 635685
Vapor Density 3.1 (vs air)
Formula Weight 92.09g/mol
Vapor Pressure 1 mmHg (20°C)
Percent Purity ≥99.5% (GC)
Grade ACS/puriss. p.a.
Chemical Name or Material Glycerol
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.