missing translation for 'onlineSavingsMsg'
Learn More

Glycerol tripalmitate, 98%

Product Code. p-7081102
Click to view available options
Quantity:
1 g
5 g
25 g
Unit Size:
1g
25g
5g
This item is not returnable. View return policy

Product Code. 11454313

Brand: Thermo Scientific Alfa Aesar A10922.03

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 555-44-2 | C51H98O6 | 807.339 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 555-44-2
Molecular Formula C51H98O6
Molecular Weight (g/mol) 807.339
MDL Number MFCD00008995
InChI Key PVNIQBQSYATKKL-UHFFFAOYSA-N
Synonym tripalmitin, glycerol tripalmitate, tripalmitoylglycerol, glyceryl tripalmitate, palmitic triglyceride, tripalmitate, triglyceryl palmitate, barolub, dynasan 116, propane-1,2,3-triyl tripalmitate
PubChem CID 11147
ChEBI CHEBI:77393
IUPAC Name 2,3-di(hexadecanoyloxy)propyl hexadecanoate
SMILES CCCCCCCCCCCCCCCC(=O)OCC(COC(=O)CCCCCCCCCCCCCCC)OC(=O)CCCCCCCCCCCCCCC

Specifications

Melting Point 60°C to 64°C
Quantity 1 g
Formula Weight 807.34
Percent Purity 98%
Chemical Name or Material Glycerol tripalmitate

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.