missing translation for 'onlineSavingsMsg'
Learn More

β-Glycerophosphate disodium salt pentahydrate, MedChemExpress

Product Code. 16422429
Encompass_Preferred
Change view
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
16422429 1 g 1g
16401198 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 16422429 Supplier Medchem Express Supplier No. HYD0886
 Restricted Item

This item requires a price update prior to being ordered.

Request a Price for this item here.

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

MedChemExpress β-Glycerophosphate disodium salt pentahydrate is a phosphatase inhibitor.

β-Glycerophosphate disodium salt pentahydrate is a phosphatase inhibitor.

Tekniske data

Chemical Name or Material β-Glycerophosphate disodium salt pentahydrate
CAS 13408-09-8
Color White
Purity Grade Notes Research
Molecular Formula C3H17Na2O11P
Quantity 1 g
Solubility Information H2O : 250 mg/mL (816.70 mM; Need ultrasonic) ∣DMSO : < 1 mg/mL (insoluble or slightly soluble)
SMILES OCC(OP(O[Na])(O[Na])=O)CO.[5H2O]
Molecular Weight (g/mol) 306.11
Formula Weight 306.11
Percent Purity 95.0%
Grade Research
Physical Form Solid
For Use With (Application) Cancer-programmed cell death
Vis mere Vis mindre

Research purposes only

Produkttitel
Vælg et problem

Ved at klikke på Send, anerkender du, at du kan blive kontaktet af Fisher Scientific med hensyn til den feedback, du har givet i denne formular. Vi deler ikke dine oplysninger til andre formål. Alle angivne kontaktoplysninger skal også vedligeholdes i overensstemmelse med vores Privatlivspolitik.