missing translation for 'onlineSavingsMsg'
Learn More

beta-Glycerophosphoric acid, disodium salt pentahydrate, 98%

Product Code. 10424701 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
25 g
100 g
500 g
Packaging:
Glass bottle
Unit Size:
100g
25g
500g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
10424701 25 g Glass bottle 25g
10259600 100 g Glass bottle 100g
10561814 500 g Glass bottle 500g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 10424701 Supplier Thermo Scientific Acros Supplier No. 410990250
Follow the Workflow - Explore the Savings
Use promo code 27967
View all promotions
 Restricted Item
Estimated Shipment: 12-11-2026
Add to basket
Edge

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

beta-Glycerophosphoric acid, disodium salt pentahydrate, 98%, C3H7Na2O6P.5H2O, CAS Number-13408-09-8 | CAS: 13408-09-8 | C3H19Na2O11P | 308.13 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 13408-09-8
Molecular Formula C3H19Na2O11P
Molecular Weight (g/mol) 308.13
MDL Number MFCD00149083,MFCD00002135,MFCD00149083,MFCD00149084,MFCD00002135
InChI Key VWVLGHQKEJESPW-UHFFFAOYSA-N
Synonym unii-5jiy5mmx0e, 5jiy5mmx0e, sodium 2-glycerophosphate pentahydrate, sodium beta-glycerophosphate pentahydrate, sodium 1,3-dihydroxypropan-2-yl phosphate pentahydrate, glycerol 2-phosphate, c3h7na2o6p.5h2o, beta-glycerophosphate, disodium salt, pentahydrate, beta-glycerol phosphate disodium salt pentahydrate, beta-glycerophosphoric acid disodium salt pentahydrate
PubChem CID 131846916
IUPAC Name 1,3-dihydroxypropan-2-yl dihydrogen phosphate;sodium;pentahydrate
SMILES O.O.O.O.O.[Na].[Na].OCC(CO)OP(O)(O)=O

Specifications

CAS Min % 97.5
CAS Max % 100.0
Melting Point 102.0°C to 104.0°C
Color White
Infrared Spectrum Authentic
Assay Percent Range 98%
Packaging Glass bottle
Linear Formula CH2OHCH(OPO3Na2)CH2OH·5H2O
Quantity 25 g
Solubility Information Solubility in water: freely soluble.
Formula Weight 306.11
Percent Purity 98%
Physical Form Crystalline Powder
Chemical Name or Material β-Glycerophosphoric acid, disodium salt pentahydrate
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.