missing translation for 'onlineSavingsMsg'
Learn More

Glyceryl Trinonadecanoate, TRC

Product Code. 30395549
Change view
Click to view available options
Quantity:
50 mg
Unit Size:
50mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30395549 50 mg 50mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30395549 Supplier Toronto Research Chemicals Supplier No. TRCG60160050MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Glyceryl Trinonadecanoate
CAS 26536-13-0
Molecular Formula C60 H116 O6
Quantity 50 mg
Synonym Glyceryl Trinonadecanoate, 1,1′,1′′-(1,2,3-Propanetriyl) trinonadecanoate (ACI), Nonadecanoic acid, 1,2,3-propanetriyl ester (9CI), Nonadecanoin, tri- (8CI), 1,2,3-Trinonadecanoylglycerol, Glyceryl trinonadecanoate, Trinonadecanoin, Nonadecanoic acid, 1,1′,1′′-(1,2,3-propanetriyl)
InChI Formula InChI=1S/C60H116O6/c1-4-7-10-13-16-19-22-25-28-31-34-37-40-43-46-49-52-58(61)64-55-57(66-60(63)54-51-48-45-42-39-36-33-30-27-24-21-18-15-12-9-6-3)56-65-59(62)53-50-47-44-41-38-35-32-29-26-23-20-17-14-11-8-5-2/h57H,4-56H2,1-3H3
SMILES CCCCCCCCCCCCCCCCCCC(=O)OCC(COC(=O)CCCCCCCCCCCCCCCCCC)OC(=O)CCCCCCCCCCCCCCCCCC
IUPAC Name 2,3-di(nonadecanoyloxy)propyl nonadecanoate
Molecular Weight (g/mol) 933.559
Formula Weight 932.877
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.