missing translation for 'onlineSavingsMsg'
Learn More

Glycidyl 2,2,3,3-tetrafluoropropyl ether, 97%, Thermo Scientific™

Product Code. 15450408
Change view
Click to view available options
Quantity:
5 g
25 g
Unit Size:
25g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15450408 25 g 25g
15440408 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 15450408 Supplier Thermo Scientific Alfa Aesar Supplier No. H53508.14
Request Bulk or Custom Format

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Applications
Glycidyl 2,2,3,3-tetrafluoropropyl ether is used as pharmaceutical intermediate.

Notes
Stable under recommended storage conditions. Incompatible with oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 19932-26-4
Molecular Formula C6H8F4O2
Molecular Weight (g/mol) 188.122
MDL Number MFCD00054682
InChI Key DATKALAKXGFGPI-UHFFFAOYSA-N
Synonym 3-2,2,3,3-tetrafluoropropoxy-1,2-epoxypropane, 2,2,3,3-tetrafluoropropoxy methyl oxirane, 2-2,2,3,3-tetrafluoropropoxy methyl oxirane, glycidyl 2,2,3,3-tetrafluoropropyl ether, 2-4,4,5,5-tetrafluoro-2-oxapentyl oxirane, oxirane, 2,2,3,3-tetrafluoropropoxy methyl, 3-2,2,3,3-tetrafluoropropoxy-1,2-propenoxide, 3-2,2,3,3,-tetrafluoropropoxy-1,2-epoxypropane, ksc497c1p
PubChem CID 89247
IUPAC Name 2-(2,2,3,3-tetrafluoropropoxymethyl)oxirane
SMILES C1C(O1)COCC(C(F)F)(F)F

Specifications

Density 1.327
Boiling Point 50°C (4 mmHg)
Flash Point 79°C (174°F)
Refractive Index 1.366
Quantity 25 g
Formula Weight 188.12
Percent Purity 97%
Chemical Name or Material Glycidyl 2,2,3,3-tetrafluoropropyl ether
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.