missing translation for 'onlineSavingsMsg'
Learn More

Glycidyl 2-methoxyphenyl ether, 98%, Thermo Scientific™

Product Code. 15448348
Change view
Click to view available options
Quantity:
5 g
25 g
Unit Size:
25g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15448348 25 g 25g
15438348 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 15448348 Supplier Thermo Scientific Alfa Aesar Supplier No. H61777.14
Request Bulk or Custom Format

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 2210-74-4
Molecular Formula C10H12O3
Molecular Weight (g/mol) 180.203
MDL Number MFCD00190158
InChI Key RJNVSQLNEALZLC-UHFFFAOYSA-N
Synonym 1-2-methoxyphenoxy-2,3-epoxypropane, guaiacol glycidyl ether, 2-2-methoxyphenoxy methyl oxirane, guajacol-glycidaether, 1,2-epoxy-3-o-methoxyphenoxy propane, glycidyl 2-methoxyphenyl ether, 1,2-epoxy-3-2-methoxyphenoxy propane, oxirane, 2-methoxyphenoxy methyl, guajacol-glycidaether german
PubChem CID 98357
IUPAC Name 2-[(2-methoxyphenoxy)methyl]oxirane
SMILES COC1=CC=CC=C1OCC2CO2

Specifications

Melting Point 41°C to 48°C
Color White
Boiling Point 152°C to 158°C
Quantity 25 g
Formula Weight 180.2
Percent Purity 98%
Physical Form Solid
Chemical Name or Material Glycidyl 2-methoxyphenyl ether
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.