missing translation for 'onlineSavingsMsg'
Learn More

Glycidyl Methacrylate, TRC

Product Code. 30390628
Change view
Click to view available options
Quantity:
500 mg
Unit Size:
500mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30390628 500 mg 500mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30390628 Supplier Toronto Research Chemicals Supplier No. TRCG650128500MG

Please to purchase this item. Need a web account? Register with us today!

This item requires a price update prior to being ordered.

Request a Price for this item here.

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material 2,3-Epoxypropyl methacrylate
CAS 106-91-2
Molecular Formula C7 H10 O3
Quantity 500 mg
Synonym 2-Propenoic acid, 2-methyl-, 2-oxiranylmethyl ester, 2-Propenoic acid, 2-methyl-, oxiranylmethyl ester (9CI), Methacrylic acid, 2,3-epoxypropyl ester (6CI,7CI,8CI), (±)-Glycidyl methacrylate, 2-Methylacrylic acid oxiranylmethyl ester, 2-[(Methacryloyloxy)methyl]oxirane, 3-Methacryloyloxy-1,2-epoxypropane, Acryester G, Acryester GMA, Acryester GU, Blemmer G, Blemmer GH, Blemmer GH-LC, Blemmer GMA, Blemmer GP, Blemmer GS, EM 54, Epoxypropyl methacrylate, Glycidol methacrylate, Glycidyl methacrylate, Glycidyl α-methylacrylate, Light Ester G, M 0590, Methacryloyloxymethyloxirane, NSC 24156, NSC 67195, P 1901, SR 379, SY-Monomer G, Sartomer 379, Sartomer SR 379
InChI Formula InChI=1S/C7H10O3/c1-5(2)7(8)10-4-6-3-9-6/h6H,1,3-4H2,2H3
SMILES CC(=C)C(=O)OCC1CO1
IUPAC Name oxiran-2-ylmethyl 2-methylprop-2-enoate
Molecular Weight (g/mol) 142.15
Formula Weight 142.063
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.