missing translation for 'onlineSavingsMsg'
Learn More

Glycidyl phenyl ether, 99%

Product Code. 11420397
Change view
Click to view available options
Quantity:
50 g
250 g
1000 g
Unit Size:
1000g
250g
50g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11420397 50 g 50g
11430397 250 g 250g
11410397 1000 g 1000g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11420397 Supplier Thermo Scientific Alfa Aesar Supplier No. B21768.18
 Restricted Item
In Stock
Add to basket
This item is not returnable. View return policy

CAS: 122-60-1 | C9H10O2 | 150.18 g/mol

2-(Phenoxymethyl)oxirane is used in the synthesis of hydrazide-containing inhibitors of HIV-1 integrase.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
2-(Phenoxymethyl)oxirane is used in the synthesis of hydrazide-containing inhibitors of HIV-1 integrase.

Solubility
Soluble in water (2.4g/L).

Notes
Store away from oxidizing agents. Keep the container tightly closed and place it in a cool, dry and well ventilated condition.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 122-60-1
Molecular Formula C9H10O2
Molecular Weight (g/mol) 150.18
MDL Number MFCD00005133,MFCD31699959
InChI Key FQYUMYWMJTYZTK-UHFFFAOYNA-N
Synonym glycidyl phenyl ether, phenyl glycidyl ether, 2-phenoxymethyl oxirane, 1,2-epoxy-3-phenoxypropane, oxirane, phenoxymethyl, phenylglycidyl ether, 2,3-epoxypropyl phenyl ether, phenoxypropene oxide, phenylglycydyl ether, ether, phenylglycidyl
PubChem CID 31217
ChEBI CHEBI:82367
IUPAC Name 2-(phenoxymethyl)oxirane
SMILES C(OC1=CC=CC=C1)C1CO1

Specifications

Melting Point 3.5°C
Density 1.11
Boiling Point 244°C to 246°C
Flash Point 114°C (237°F)
Odor Sweet
Refractive Index 1.531
Quantity 50 g
Beilstein 2744
Solubility Information Soluble in water (2.4g/L).
Formula Weight 150.18
Percent Purity 99%
Chemical Name or Material Glycidyl phenyl ether
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.