missing translation for 'onlineSavingsMsg'
Learn More

Glycine, MP Biomedicals™

Product Code. 11424966 Shop All MP Biomedicals Products
Change view
Click to view available options
:
Molecular weight: 75.067 g/mol
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code.
11424966 Molecular weight: 75.067 g/mol
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 11424966 Supplier MP Biomedicals Supplier No. 04808873

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Component of Tris-glycine and Tris-glycine-SDS running buffers for polyacrylamide gel electrophoresis and Towbin's transfer buffer for western blots.

  • Glycine is a component of Tris-glycine and Tris-glycine-SDS running buffers for polyacrylamide gel electrophoresis.
  • Glycine is also a component of Towbin's transfer buffer for western blots.
  • Research Use Only (RUO). Ready for CE IVD labeling of clinical applications.
  • Key applications: Electrophoresis, SDS PAGE.
  • Inhibitory neurotransmitter in spinal cord, allosteric regulator of NMDA receptors.
TRUSTED_SUSTAINABILITY

Specifications

Melting Point 451°F (NTP, 1992)
CAS 56-40-6
pH 4.0 (0.2 M solution in water)
Color White
Heavy Metals ≤20 ppm
Loss on Drying ≤0.2%
Molecular Formula C2H5NO2
MDL Number MFCD00008131
Beilstein 635782
Synonym Aminoacetic acid, Gly, Aminoethanoic acid, Glycocoll
Solubility Information ≥ 100 mg/mL at 64°F (NTP, 1992)
Density 1.1607 (NTP, 1992)
Molecular Weight (g/mol) 75.067
Optical Rotation ≤0.0065%
Percent Purity ≥99.5%
Grade Electrophoresis
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.