missing translation for 'onlineSavingsMsg'
Learn More

Glycine 6-Ethylchenodeoxycholate, TRC

Product Code. 30138288
Change view
Click to view available options
Quantity:
10 mg
25 mg
50 mg
Unit Size:
10mg
25mg
50mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30138288 50 mg 50mg
30137245 25 mg 25mg
30003246 10 mg 10mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30138288 Supplier Toronto Research Chemicals Supplier No. TRCG62525050MG
 Restricted Item
Estimated Shipment: 14-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 863239-60-5
Molecular Formula C28H47NO5
Molecular Weight (g/mol) 477.68
Synonym N-[(3α,5β,6α,7α)-6-Ethyl-3,7-dihydroxy-24-oxocholan-24-yl]glycine, Glyco Obeticholic Acid;
IUPAC Name 2-[[(4R)-4-[(3R,5S,6R,7R,8S,9S,10S,13R,14S,17R)-6-ethyl-3,7-dihydroxy-10,13-dimethyl-2,3,4,5,6,7,8,9,11,12,14,15,16,17-tetradecahydro-1H-cyclopenta[a]phenanthren-17-yl]pentanoyl]amino]acetic acid
SMILES C[C@@]12[C@](CC[C@]2([H])[C@H](C)CCC(NCC(O)=O)=O)([H])[C@]3([H])[C@H](O)[C@H](CC)[C@@]4([H])[C@@](CC[C@@H](O)C4)(C)[C@@]3([H])CC1

Specifications

InChI Formula InChI=1S/C28H47NO5/c1-5-18-22-14-17(30)10-12-28(22,4)21-11-13-27(3)19(7-8-20(27)25(21)26(18)34)16(2)6-9-23(31)29-15-24(32)33/h16-22,25-26,30,34H,5-15H2,1-4H3,(H,29,31)(H,32,33)/t16-,17-,18-,19-,20+,21+,22+,25+,26-,27-,28-/m1/s1
Quantity 50 mg
Formula Weight 477.3454
Percent Purity >95%
Shipping Condition Room Temperature
Chemical Name or Material Glycine 6-Ethylchenodeoxycholate
Grade HPLC
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.