missing translation for 'onlineSavingsMsg'
Learn More

Glycine Anhydride (2,5-Piperazinedione), TRC

Product Code. 30138370
Change view
Click to view available options
Quantity:
1 g
10 g
Unit Size:
10g
1g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30138370 10 g 10g
17947534 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30138370 Supplier Toronto Research Chemicals Supplier No. TRCP47992010G
 Restricted Item
Estimated Shipment: 01-09-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 106-57-0
Molecular Formula C4 H6 N2 O2
Molecular Weight (g/mol) 114.1
Synonym 2,5-Piperazinedione, 2,5-Diketopiperazine, 2,5-Dioxopiperazine, Cyclo(Gly-Gly), Cyclo(glycylglycyl), Cyclodiglycine, Cycloglycylglycine, Diglycolyl diamide, Diketopiperazine, Glycine cyclic dimer, Glycine, N-glycyl-, cyclic peptide, Glycine, bimol. cyclic peptide, Glycylglycine lactam, NSC 26345, α,γ-Diacipiperazine
IUPAC Name piperazine-2,5-dione
SMILES O=C1CNC(=O)CN1

Specifications

InChI Formula InChI=1S/C4H6N2O2/c7-3-1-5-4(8)2-6-3/h1-2H2,(H,5,8)(H,6,7)
Quantity 10 g
Formula Weight 114.0429
Percent Purity >95%
Shipping Condition Room Temperature
Chemical Name or Material Glycine Anhydride (2,5-Piperazinedione)
Grade HPLC
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.