missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals Glycine anhydride, 98%

Product Code. 11483098
Change view
Click to view available options
Quantity:
25 g
100 g
Unit Size:
100g
25g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11483098 100 g 100g
11473098 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11483098 Supplier Thermo Scientific Chemicals Supplier No. A18822.22
Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 106-57-0 | C4H6N2O2 | 114.104 g/mol

It can be used as an intermediate in the preparation of various pharmaceutical and biologically active compounds. Also used in the synthesis of dipeptide isosteres by cross-metathesis.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It can be used as an intermediate in the preparation of various pharmaceutical and biologically active compounds. Also used in the synthesis of dipeptide isosteres by cross-metathesis.

Solubility
Soluble in water (142mg/L).

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from oxidizing agent.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 106-57-0
Molecular Formula C4H6N2O2
Molecular Weight (g/mol) 114.104
MDL Number MFCD00006009
InChI Key BXRNXXXXHLBUKK-UHFFFAOYSA-N
Synonym 2,5-piperazinedione, 2,5-dioxopiperazine, 2,5-diketopiperazine, cyclodiglycine, cycloglycylglycine, cyclo glycylglycyl, glycylglycine lactam, diglycolyl diamide, glycine, bimol. cyclic peptide, alpha,gamma-diacipiperazine
PubChem CID 7817
ChEBI CHEBI:16535
IUPAC Name piperazine-2,5-dione
SMILES C1C(=O)NCC(=O)N1

Specifications

Melting Point ∼300°C (decomposition)
Beilstein 112112
Merck Index 14,7466
Solubility Information Soluble in water (142mg/L).
Quantity 100 g
Formula Weight 114.1
Percent Purity 99%
Chemical Name or Material Glycine anhydride

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.