missing translation for 'onlineSavingsMsg'
Learn More

Glycine anhydride, 98%

Product Code. 10764801 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
1 kg
Packaging:
Glass bottle
Unit Size:
100g
1kg
25g
5g
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
10764801 100 g Glass bottle 100g
10393952 5 g Glass bottle 5g
10754801 25 g Glass bottle 25g
10513314 1 kg Glass bottle 1kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
This item is not returnable. View return policy
Product Code. 10764801 Supplier Thermo Scientific Acros Supplier No. 120081000
 Restricted Item
Estimated Shipment: 11-09-2026
Add to basket
This item is not returnable. View return policy

CAS: 106-57-0 | C4H6N2O2 | 114.1 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 106-57-0
Molecular Formula C4H6N2O2
Molecular Weight (g/mol) 114.1
MDL Number MFCD00006009
InChI Key BXRNXXXXHLBUKK-UHFFFAOYSA-N
Synonym 2,5-piperazinedione, 2,5-dioxopiperazine, 2,5-diketopiperazine, cyclodiglycine, cycloglycylglycine, cyclo glycylglycyl, glycylglycine lactam, diglycolyl diamide, glycine, bimol. cyclic peptide, alpha,gamma-diacipiperazine
PubChem CID 7817
ChEBI CHEBI:16535
IUPAC Name piperazine-2,5-dione
SMILES C1C(=O)NCC(=O)N1

Specifications

Melting Point >300.0°C
Color White to Yellow
Loss on Drying 0.5% max. (105°C, 4 hrs)
Infrared Spectrum Authentic
Assay Percent Range 97.5% min. (ex N)
Packaging Glass bottle
Quantity 100 g
Beilstein 24, 264
Merck Index 15, 7578
Solubility Information Solubility in water: 142g/L water (20 c). Other solubilities: soluble in hcl, slightly soluble in ethanol
Formula Weight 114.1
Percent Purity 98%
Physical Form Crystalline Powder
Chemical Name or Material Glycine anhydride, 98%
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.