missing translation for 'onlineSavingsMsg'
Learn More

Glycine dimethylamide, 97%

Product Code. 11449440
Change view
Click to view available options
Quantity:
250 mg
1 g
5 g
Unit Size:
1g
250mg
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11449440 1 g 1g
11439440 250 mg 250mg
15457867 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11449440 Supplier Thermo Scientific Alfa Aesar Supplier No. H50293.03
 Restricted Item
Estimated Shipment: 19-10-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 1857-19-8 | C4H10N2O | 102.137 g/mol

Glycine dimethylamide is used as pharmaceutical intermediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Glycine dimethylamide is used as pharmaceutical intermediate.

Notes
Air sensitive. Incompatible with oxidizing agents and acids.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1857-19-8
MDL Number MFCD06655265
Synonym 2-amino-n,n-dimethyl-acetamide, acetamide, 2-amino-n,n-dimethyl, n,n-dimethylglycinamide, glycinedimethyl amide, pubchem22320, acmc-1bw2m, 1,1-dimethylurea hydrochloride, 2-amino-n,n,-dimethylacetamide, 2-amino-n,n-dimethylacetamide; n,n-dimethylglycinamide
PubChem CID 4961823

Specifications

Molecular Formula C4H10N2O
Linear Formula H2NCH2CON(CH3)2
UN Number UN2735
InChI Key KNVRBEGQERGQRP-UHFFFAOYSA-N
SMILES CN(C)C(=O)CN
IUPAC Name 2-amino-N,N-dimethylacetamide
Molecular Weight (g/mol) 102.137
Quantity 1 g
Odor Amine-like
Formula Weight 102.14
Sensitivity Air Sensitive
Percent Purity 97%
Chemical Name or Material Glycine dimethylamide
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.