missing translation for 'onlineSavingsMsg'
Learn More

Glycine, Electrophoresis Grade, MP Biomedicals™

Product Code. 11491642 Shop All MP Biomedicals Products
Click to view available options
Quantity:
1 kg
5 kg
Unit Size:
1kg
5kg
This item is not returnable. View return policy

Product Code. 11491642

Brand: MP Biomedicals 04808831

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Glycine, Electrophoresis Grade is for use in electrophoresis buffers and SDS-PAGE.

  • Glycine is a component of Tris-glycine and Tris-glycine-SDS running buffers for polyacrylamide gel electrophoresis.
  • Glycine is also a component of Towbin′s transfer buffer for Western blots.
  • It is an inhibitory neurotransmitter in the spinal cord and an allosteric regulator of NMDA receptors.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 56-40-6
Molecular Formula C2H5NO2
Molecular Weight (g/mol) 75.07
MDL Number MFCD00008131
InChI Key DHMQDGOQFOQNFH-UHFFFAOYSA-N
Synonym glycine, aminoacetic acid, glycocoll, aminoethanoic acid, glycolixir, glycosthene, aciport, glicoamin, padil, hampshire glycine
PubChem CID 750
ChEBI CHEBI:15428
IUPAC Name 2-aminoacetic acid
SMILES NCC(O)=O

Specifications

Melting Point 451°F
pH 4 (0.2 molar aq. soln.)
Specific Rotation ≤0.0065%
Solubility Information Soluble in water. Slightly soluble in alcohol and ethanol (95%). Almost insoluble in ether.
Density 1.1607 g/mL
Quantity 5 kg
Optical Rotation ≤0.0065%
Percent Purity ≥99.5%
Grade Electrophoresis
Physical Form Powder
pKa 2.35 at 25°C
Chemical Name or Material Glycine
Show More Show Less

Unless specified otherwise, MP Biomedical's products are for research or further manufacturing use only, not for direct human use.

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

Indem Sie auf Absenden klicken, erklären Sie sich damit einverstanden, dass Fisher Scientific sich mit Ihnen in Verbindung setzen kann, um Ihr Feedback in diesem Formular zu bearbeiten. Wir werden Ihre Informationen nicht für andere Zwecke weitergeben. Alle bereitgestellten Kontaktinformationen werden in Übereinstimmung mit unserer Datenschutzrichtlinie aufbewahrt. Datenschutzrichtlinie.

Vielen Dank, dass Sie uns helfen, unsere Website zu verbessern. Ihr Feedback wurde übermittelt