missing translation for 'onlineSavingsMsg'
Learn More

Glycine, Ultrapure Bioreagent, J.T. Baker™

Product Code. p-5383268 Shop All J.T.Baker Products
Click to view available options
Quantity:
250 g
1 kg
5 kg
Packaging:
Poly Bottle
Poly Pail
Unit Size:
Each
This item is not returnable. View return policy

Product Code. 15518234

Brand: Avantor J.T.Baker 405900

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High purity reagents tested for use in biotechnology applications, such as electrophoresis, and liquid chromatography

    For Electrophoresis, Liquid Chromatography and Molecular Biology Applications

Chemical Identifiers

CAS 56-40-6
Molecular Formula C2H5NO2
Molecular Weight (g/mol) 75.07
MDL Number MFCD00008131
InChI Key DHMQDGOQFOQNFH-UHFFFAOYSA-N
Synonym glycine, aminoacetic acid, glycocoll, aminoethanoic acid, glycolixir, glycosthene, aciport, glicoamin, padil, hampshire glycine
PubChem CID 750
ChEBI CHEBI:15428
IUPAC Name 2-aminoacetic acid
SMILES NCC(O)=O

Specifications

Packaging Poly Bottle
Quantity 250 g
Formula Weight 75.07
Specific Gravity 1L = 1.16kg
Grade Ultrapure Bioreagent
Chemical Name or Material Glycine
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.