missing translation for 'onlineSavingsMsg'
Learn More

Glycochenodeoxycholic Acid 3-Sulfate Disodium Salt, TRC

Product Code. 30007303
Change view
Click to view available options
Quantity:
2.5 mg
25 mg
Unit Size:
2.5mg
25mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30007303 2.5 mg 2.5mg
30005625 25 mg 25mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30007303 Supplier Toronto Research Chemicals Supplier No. TRCG6412702.5MG
 Restricted Item
Estimated Shipment: 01-09-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 66874-09-7
Molecular Formula C26H41NNa2O8S
Molecular Weight (g/mol) 573.65
Synonym N-[(3α,5β,7α)-7-Hydroxy-24-oxo-3-(sulfooxy)cholan-24-yl]glycine Disodium Salt, Glycochenodeoxycholic Acid 3α-Sulfate Disodium Salt;
SMILES C[C@H](CCC(=O)NCC(=O)[O-])[C@H]1CC[C@@H]2[C@@]1(CC[C@H]3[C@H]2[C@@H](C[C@H]4[C@@]3(CC[C@H](C4)OS(=O)(=O)[O-])C)O)C.[Na+].[Na+]

Specifications

Quantity 2.5 mg
InChI Formula InChI=1S/C26H43NO8S.2Na/c1-15(4-7-22(29)27-14-23(30)31)18-5-6-19-24-20(9-11-26(18,19)3)25(2)10-8-17(35-36(32,33)34)12-16(25)13-21(24)28;;/h15-21,24,28H,4-14H2,1-3H3,(H,27,29)(H,30,31)(H,32,33,34);;/q;2*+1/p-2/t15-,16+,17-,18-,19+,20+,21-,24+,25+,26-;;/m1../s1
Formula Weight 573.2348
Percent Purity >95%
Chemical Name or Material Glycochenodeoxycholic Acid 3-Sulfate Disodium Salt
Grade HPLC
Produkttitel
Vælg et problem

Ved at klikke på Send, anerkender du, at du kan blive kontaktet af Fisher Scientific med hensyn til den feedback, du har givet i denne formular. Vi deler ikke dine oplysninger til andre formål. Alle angivne kontaktoplysninger skal også vedligeholdes i overensstemmelse med vores Privatlivspolitik.