missing translation for 'onlineSavingsMsg'
Learn More

Glycochenodeoxycholic Acid Sodium Salt (Ultra pure, >97%), TRC

Product Code. 30240785
Change view
Click to view available options
Quantity:
50 mg
250 mg
1 g
Unit Size:
1g
250mg
50mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30240785 50 mg 50mg
30240646 1 g 1g
30240669 250 mg 250mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30240785 Supplier Toronto Research Chemicals Supplier No. TRCG64126050MG
 Restricted Item
Estimated Shipment: 01-09-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 16564-43-5
Molecular Formula C26 H42 N O5 . Na
Molecular Weight (g/mol) 471.61
Synonym N-[(3alpha,5beta,7alpha)-3,7-Dihydroxy-24-oxocholan-24-yl]glycine Sodium Salt, N-[(3alpha,5beta,7alpha)-3,7-Dihydroxy-24-oxocholan-24-yl]glycine Monosodium Salt, NSC 681056, Sodium Chenodeoxycholylglycine, Sodium Glycochenodeoxycholate, Sodium Glycochenodesoxycholate;
IUPAC Name sodium;2-[[(4R)-4-[(3R,5S,7R,8R,9S,10S,13R,14S,17R)-3,7-dihydroxy-10,13-dimethyl-2,3,4,5,6,7,8,9,11,12,14,15,16,17-tetradecahydro-1H-cyclopenta[a]phenanthren-17-yl]pentanoyl]amino]acetate
SMILES [Na+].C[C@H](CCC(=O)NCC(=O)[O-])[C@H]1CC[C@H]2[C@@H]3[C@H](O)C[C@@H]4C[C@H](O)CC[C@]4(C)[C@H]3CC[C@]12C

Specifications

Quantity 50 mg
InChI Formula InChI=1S/C26H43NO5.Na/c1-15(4-7-22(30)27-14-23(31)32)18-5-6-19-24-20(9-11-26(18,19)3)25(2)10-8-17(28)12-16(25)13-21(24)29;/h15-21,24,28-29H,4-14H2,1-3H3,(H,27,30)(H,31,32);/q;+1/p-1/t15-,16+,17-,18-,19+,20+,21-,24+,25+,26-;/m1./s1
Formula Weight 471.2961
Percent Purity >95%
Chemical Name or Material Glycochenodeoxycholic Acid Sodium Salt
Grade HPLC
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.