missing translation for 'onlineSavingsMsg'
Learn More

Glycocholic Acid Hydrate Sodium Salt, TRC

Product Code. 30010903
Change view
Click to view available options
Quantity:
5 g
25 g
50 g
Unit Size:
25g
50g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30010903 5 g 5g
30006137 25 g 25g
30009701 50 g 50g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30010903 Supplier Toronto Research Chemicals Supplier No. TRCG6413605G
 Restricted Item
Estimated Shipment: 15-10-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 338950-81-5
Molecular Formula C26 H42 N O6 . Na . H2 O
Molecular Weight (g/mol) 505.62
Synonym Glycine, N-[(3α,5β,7α,12α)-3,7,12-trihydroxy-24-oxocholan-24-yl]-, monosodium salt, hydrate (9CI), Glycocholic acid sodium salt hydrate, Sodium glycocholate hydrate, N-[(3α,5β,7α,12α)-3,7,12-Trihydroxy-24-oxocholan-24-yl]glycine sodium salt hydrate, Glycocholic acid sodium salt hydrate, Monosodium glycocholate hydrate, Sodium cholylglycinate hydrate
IUPAC Name sodium;2-[[(4R)-4-[(3R,5S,7R,8R,9S,10S,12S,13R,14S,17R)-3,7,12-trihydroxy-10,13-dimethyl-2,3,4,5,6,7,8,9,11,12,14,15,16,17-tetradecahydro-1H-cyclopenta[a]phenanthren-17-yl]pentanoyl]amino]acetate;hydrate
SMILES O.[Na+].C[C@H](CCC(=O)NCC(=O)[O-])[C@H]1CC[C@H]2[C@@H]3[C@H](O)C[C@@H]4C[C@H](O)CC[C@]4(C)[C@H]3C[C@H](O)[C@]12C

Specifications

Quantity 5 g
InChI Formula InChI=1S/C26H43NO6.Na.H2O/c1-14(4-7-22(31)27-13-23(32)33)17-5-6-18-24-19(12-21(30)26(17,18)3)25(2)9-8-16(28)10-15(25)11-20(24)29;;/h14-21,24,28-30H,4-13H2,1-3H3,(H,27,31)(H,32,33);;1H2/q;+1;/p-1/t14-,15+,16-,17-,18+,19+,20-,21+,24+,25+,26-;;/m1../s1
Formula Weight 505.302
Chemical Name or Material Glycocholic Acid Hydrate Sodium Salt
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.