missing translation for 'onlineSavingsMsg'
Learn More

Glycocholic acid hydrate, Thermo Scientific™

Product Code. 17221316 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
1 g
5 g
Packaging:
Glass Bottle
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
17221316 5 g Glass Bottle 5g
17211316 1 g Glass Bottle 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 17221316 Supplier Thermo Scientific Acros Supplier No. 466790050
 Restricted Item
Estimated Shipment: 25-08-2026
Add to basket
This item is not returnable. View return policy
This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1192657-83-2
Molecular Formula C26H43NO6
Molecular Weight (g/mol) 465.63
MDL Number MFCD06408004
InChI Key RFDAIACWWDREDC-MTSIQNSJNA-N
IUPAC Name 2-(4-{4,7,11-trihydroxy-9a,11a-dimethyl-hexadecahydro-1H-cyclopenta[a]phenanthren-1-yl}pentanamido)acetic acid hydrate
SMILES [H][C@@]1(CC[C@@]2([H])[C@]3([H])[C@H](O)C[C@]4([H])C[C@H](O)CC[C@]4(C)[C@@]3([H])C[C@H](O)[C@]12C)[C@H](C)CCC(=O)NCC(O)=O

Specifications

Alpha Vector Glycocholicacid
Color White to Yellow
Infrared Spectrum Conforms
Packaging Glass Bottle
Quantity 5 g
Solubility Information (50 mg/ml Ethanol) Clear colorless to light yellow solution
Formula Weight 486.64
Physical Form Powder
Chemical Name or Material Glycocholic acid hydrate
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.