missing translation for 'onlineSavingsMsg'
Learn More

Glycocholic Acid Hydrate, TRC

Product Code. 30135389
Change view
Click to view available options
Quantity:
1 g
10 g
25 g
Unit Size:
10g
1g
25g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30135389 25 g 25g
30134780 10 g 10g
30004091 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30135389 Supplier Toronto Research Chemicals Supplier No. TRCG64135025G
 Restricted Item
Estimated Shipment: 15-10-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1192657-83-2
Molecular Formula C26 H43 N O6 . x[H2 O]
Molecular Weight (g/mol) 465.52
Synonym N-[(3α,5β,7α,12α)-3,7,12-Trihydroxy-24-oxocholan-24-yl]glycine Hydrate, N-Choloylglycine-d4, Glycoreductodehydrocholic Acid Hydrate, 3α,7α,12α-Trihydroxy-5β-cholanic αcid-24-glycine Hydrate, Cholylglycine Hydrate, Glycine Cholate Hydrate;
IUPAC Name 2-[[(4R)-4-[(3R,5S,7R,8R,9S,10S,12S,13R,14S,17R)-3,7,12-trihydroxy-10,13-dimethyl-2,3,4,5,6,7,8,9,11,12,14,15,16,17-tetradecahydro-1H-cyclopenta[a]phenanthren-17-yl]pentanoyl]amino]acetic acid;hydrate
SMILES O.C[C@H](CCC(=O)NCC(=O)O)[C@H]1CC[C@H]2[C@@H]3[C@H](O)C[C@@H]4C[C@H](O)CC[C@]4(C)[C@H]3C[C@H](O)[C@]12C

Specifications

Quantity 25 g
InChI Formula InChI=1S/C26H43NO6.H2O/c1-14(4-7-22(31)27-13-23(32)33)17-5-6-18-24-19(12-21(30)26(17,18)3)25(2)9-8-16(28)10-15(25)11-20(24)29;/h14-21,24,28-30H,4-13H2,1-3H3,(H,27,31)(H,32,33);1H2/t14-,15+,16-,17-,18+,19+,20-,21+,24+,25+,26-;/m1./s1
Formula Weight 483.3196
Percent Purity >95%
Chemical Name or Material Glycocholic Acid Hydrate
Grade HPLC
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.