missing translation for 'onlineSavingsMsg'
Learn More

Glycocholic Acid Sodium Salt, TRC

Product Code. 30007358
Change view
Click to view available options
Quantity:
100 mg
500 mg
1 g
Unit Size:
100mg
1g
500mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30007358 1 g 1g
30003302 500 mg 500mg
30008010 100 mg 100mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30007358 Supplier Toronto Research Chemicals Supplier No. TRCG6413681G
 Restricted Item
Estimated Shipment: 14-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 863-57-0
Molecular Formula C26 H42 N O6 . Na
Molecular Weight (g/mol) 487.6045
Synonym Glycine, N-[(3α,5β,7α,12α)-3,7,12-trihydroxy-24-oxocholan-24-yl]-, sodium salt (1:1), Glycine, N-[(3α,5β,7α,12α)-3,7,12-trihydroxy-24-oxocholan-24-yl]-, monosodium salt (9CI), Glycine, N-choloyl-, monosodium salt (8CI), Cholane, glycine deriv., Monosodium glycocholate, NSC 35609, Sodium cholylglycinate, Sodium glycocholate, N-[(3α,5β,7α,12α)-3,7,12-Trihydroxy-24-oxocholan-24-yl]glycine Sodium Salt, N-Choloylglycine Sodium Salt
IUPAC Name sodium;2-[[(4R)-4-[(3R,5S,7R,8R,9S,10S,12S,13R,14S,17R)-3,7,12-trihydroxy-10,13-dimethyl-2,3,4,5,6,7,8,9,11,12,14,15,16,17-tetradecahydro-1H-cyclopenta[a]phenanthren-17-yl]pentanoyl]amino]acetate
SMILES [Na+].C[C@H](CCC(=O)NCC(=O)[O-])[C@H]1CC[C@H]2[C@@H]3[C@H](O)C[C@@H]4C[C@H](O)CC[C@]4(C)[C@H]3C[C@H](O)[C@]12C

Specifications

Quantity 1 g
InChI Formula InChI=1S/C26H43NO6.Na/c1-14(4-7-22(31)27-13-23(32)33)17-5-6-18-24-19(12-21(30)26(17,18)3)25(2)9-8-16(28)10-15(25)11-20(24)29;/h14-21,24,28-30H,4-13H2,1-3H3,(H,27,31)(H,32,33);/q;+1/p-1/t14-,15+,16-,17-,18+,19+,20-,21+,24+,25+,26-;/m1./s1
Formula Weight 487.291
Chemical Name or Material Glycocholic Acid Sodium Salt
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.