missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals Glycogen, From Oyster

Product Code. 10065690 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
1 g
5 g
Packaging:
Glass bottle
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10065690 1 g Glass bottle 1g
10015273 5 g Glass bottle 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Dieser Artikel kann nicht zurückgegeben werden. Rückgaberichtlinie anzeigen
Artikelnummer. 10065690 Lieferant Thermo Scientific Chemicals Lieferanten-Nr. 422950010
Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

Dieser Artikel kann nicht zurückgegeben werden. Rückgaberichtlinie anzeigen

Glycogen, CAS # 9005-79-2 is a highly branched, polysaccharide form of glucose used for energy storage in humans and other animals. | CAS: 9005-79-2 | C24H42O21 | 666.579 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

General Description

  • Glycogen is the main form in which energy is stored in animals as well as other higher organisms
  • It consists of a core protein unit surrounded by highly branched glucose units
  • Glycogen is synthesized through a series of enzymatic reactions and stored mainly in the liver and skeletal muscle to provide energy during fasting periods and during muscle contraction
  • Glycogen is an analog of starch, an energy store in plants, though it is both more compact and more highly branched than starch

Applications

  • Glycogen is most commonly used as an inert carrier substance for nucleic acid precipitation that may increase DNA and RNA extraction efficiency
TRUSTED_SUSTAINABILITY

Chemische Identifikatoren

CAS 9005-79-2
Molecular Formula C24H42O21
Molecular Weight (g/mol) 666.579
MDL Number MFCD00081547
InChI Key BYSGBSNPRWKUQH-UJDJLXLFSA-N
Synonym glycogen, 2r,3r,4s,5s,6r-2-2r,3s,4r,5r,6r-4,5-dihydroxy-6-2r,3s,4r,5r,6s-4,5,6-trihydroxy-2-hydroxymethyl oxan-3-yl oxy-2-2s,3r,4s,5s,6r-3,4,5-trihydroxy-6-hydroxymethyl oxan-2-yl oxy methyl oxan-3-yl oxy-6-hydroxymethyl oxane-3,4,5-triol, glycogen from bovine liver, 2r,3r,4s,5s,6r-2-2r,3s,4r,5r,6r-4,5-dihydroxy-6-2r,3s,4r,5r,6s-4,5,6-trihydroxy-2-hydroxymethyl oxan-3-yl oxy-2-2s,3r,4s,5s,6r-3,4,5-trihydroxy-6-hydroxymethyl oxan-2-yl oxymethyl oxan-3-yl oxy-6-hydroxymethyl oxane-3,4,5-triol, 2s,3r,4s,5s,6r-2-2r,3s,4r,5r,6r-4,5-dihydroxy-3-2r,3r,4s,5s,6r-3,4,5-trihydroxy-6-hydroxymethyl oxan-2-yl oxy-6-2r,3s,4r,5r,6s-4,5,6-trihydroxy-2-hydroxymethyl oxan-3-yl oxyoxan-2-yl methoxy-6-hydroxymethyl oxane-3,4,5-triol, 4-o-4-o-alpha-d-glucopyranosyl-6-o-alpha-d-glucopyranosyl-alpha-d-glucopyranosyl-alpha-d-glucopyranose
PubChem CID 439177
ChEBI CHEBI:28087
IUPAC Name (2S,3R,4S,5S,6R)-2-[[(2R,3S,4R,5R,6R)-4,5-dihydroxy-3-[(2R,3R,4S,5S,6R)-3,4,5-trihydroxy-6-(hydroxymethyl)oxan-2-yl]oxy-6-[(2R,3S,4R,5R,6S)-4,5,6-trihydroxy-2-(hydroxymethyl)oxan-3-yl]oxyoxan-2-yl]methoxy]-6-(hydroxymethyl)oxane-3,4,5-triol
SMILES C(C1C(C(C(C(O1)OCC2C(C(C(C(O2)OC3C(OC(C(C3O)O)O)CO)O)O)OC4C(C(C(C(O4)CO)O)O)O)O)O)O)O

Spezifikation

Color White to Beige
Loss on Drying 10% max. (105°C, 3 hrs)
Infrared Spectrum Authentic
Packaging Glass bottle
Linear Formula (C6H10O5)n
Quantity 1 g
Merck Index 15, 4531
Specific Rotation Condition +175° (20°C c=0.1,water)
Specific Rotation +160.0° to +180.0° (20°C, 589nm) (c=0.1, water)
Physical Form Powder
Chemical Name or Material Glycogen, From oyster
Mehr anzeigen Weniger anzeigen

RUO – Research Use Only

Name des Produkts
Wählen Sie ein Thema

Indem Sie auf Absenden klicken, erklären Sie sich damit einverstanden, dass Fisher Scientific sich mit Ihnen in Verbindung setzen kann, um Ihr Feedback in diesem Formular zu bearbeiten. Wir werden Ihre Informationen nicht für andere Zwecke weitergeben. Alle bereitgestellten Kontaktinformationen werden in Übereinstimmung mit unserer Datenschutzrichtlinie aufbewahrt. Datenschutzrichtlinie.