missing translation for 'onlineSavingsMsg'
Learn More

Glycogen, from Oysters, Ultrapure

Product Code. 15825158 Shop All Thermo Scientific Products
Change view
Click to view available options
Quantity:
5 g
100 g
Packaging:
Plastic bottle
Unit Size:
100g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
15825158 5 g Plastic bottle 5g
15782808 100 g Plastic bottle 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 15825158 Supplier Thermo Scientific Supplier No. J16445.06
Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

CAS: 9005-79-2 | C24H42O21 | 666.579 g/mol

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 9005-79-2
Molecular Formula C24H42O21
Molecular Weight (g/mol) 666.579
MDL Number MFCD00081547
InChI Key BYSGBSNPRWKUQH-UJDJLXLFSA-N
Synonym glycogen, 2 r,3 r,4 s,5 s,6 r-2-2 r,3 s,4 r,5 r,6 r-4,5-dihydroxy-6-2 r,3 s,4 r,5 r,6 s-4,5,6-trihydroxy-2-hydroxymethyl oxan-3-yl oxy-2-2 s,3 r,4 s,5 s,6 r-3,4,5-trihydroxy-6-hydroxymethyl oxan-2-yl oxy methyl oxan-3-yl oxy-6-hydroxymethyl oxane-3,4,5-triol, glycogen from bovine liver, 2 r,3 r,4 s,5 s,6 r-2-2 r,3 s,4 r,5 r,6 r-4,5-dihydroxy-6-2 r,3 s,4 r,5 r,6 s-4,5,6-trihydroxy-2-hydroxymethyl oxan-3-yl oxy-2-2 s,3 r,4 s,5 s,6 r-3,4,5-trihydroxy-6-hydroxymethyl oxan-2-yl oxymethyl oxan-3-yl oxy-6-hydroxymethyl oxane-3,4,5-triol, 2 s,3 r,4 s,5 s,6 r-2-2 r,3 s,4 r,5 r,6 r-4,5-dihydroxy-3-2 r,3 r,4 s,5 s,6 r-3,4,5-trihydroxy-6-hydroxymethyl oxan-2-yl oxy-6-2 r,3 s,4 r,5 r,6 s-4,5,6-trihydroxy-2-hydroxymethyl oxan-3-yl oxyoxan-2-yl methoxy-6-hydroxymethyl oxane-3,4,5-triol, 4-o-4-o-alpha-d-glucopyranosyl-6-o-alpha-d-glucopyranosyl-alpha-d-glucopyranosyl-alpha-d-glucopyranose
PubChem CID 439177
ChEBI CHEBI:28087
IUPAC Name (2 S,3 R,4 S,5 S,6 R)-2-[[(2 R,3 S,4 R,5 R,6 R)-4,5-dihydroxy-3-[(2 R,3 R,4 S,5 S,6 R)-3,4,5-trihydroxy-6-(hydroxymethyl)oxan-2-yl]oxy-6-[(2 R,3 S,4 R,5 R,6 S)-4,5,6-trihydroxy-2-(hydroxymethyl)oxan-3-yl]oxyoxan-2-yl]methoxy]-6-(hydroxymethyl)oxane-3,4,5-triol
SMILES C(C1C(C(C(C(O1)OCC2C(C(C(C(O2)OC3C(OC(C(C3O)O)O)CO)O)O)OC4C(C(C(C(O4)CO)O)O)O)O)O)O)O

Specifications

Packaging Plastic bottle
Quantity 5 g
Grade Ultrapure
Physical Form Powder
Chemical Name or Material Glycogen

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.