missing translation for 'onlineSavingsMsg'
Learn More

Glycol sulfite, 98%

Product Code. 15456608
Change view
Click to view available options
Quantity:
5 g
25 g
Unit Size:
25g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15456608 25 g 25g
15446608 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 15456608 Supplier Thermo Scientific Alfa Aesar Supplier No. H60133.14
Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 3741-38-6 | C2H4O3S | 108.11 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 3741-38-6
Molecular Formula C2H4O3S
Molecular Weight (g/mol) 108.11
MDL Number MFCD00005354
InChI Key WDXYVJKNSMILOQ-UHFFFAOYSA-N
Synonym ethylene sulfite, glycol sulfite, 1,3,2-dioxathiolane, 2-oxide, glycolsulfite, 1,2-ethylene sulfite, cyclic ethylene sulfite, ethylene glycol cyclic sulfite, ethylene glycol, cyclic sulfite, ethylene sulphite, sulfurous acid, cyclic ester with ethylene glycol
PubChem CID 77342
IUPAC Name 1,3,2-dioxathiolane 2-oxide
SMILES O=S1OCCO1

Specifications

Density 1.426
Boiling Point 159°C
Flash Point 79°C (174°F)
Refractive Index 1.445
Quantity 25 g
Beilstein 1237109
Formula Weight 108.12
Percent Purity 98%
Chemical Name or Material Glycol sulfite

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.