missing translation for 'onlineSavingsMsg'
Learn More

Glycolic acid, 98%

Product Code. 11424164
Change view
Click to view available options
Quantity:
100 g
500 g
Unit Size:
100g
500g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11424164 100 g 100g
11434164 500 g 500g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11424164 Supplier Thermo Scientific Alfa Aesar Supplier No. A12511.22
Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 79-14-1 | C2H4O3 | 76.051 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Suitable for battery materials development.

Applications
Glycolic acid is used for acne and anti-aging treatment. It is also employed in the textile industry as a dyeing and tanning agent. It is used in emulsion polymers, solvents and additives for ink and paints. It is also useful as an intermediate for organic synthesis such as redox reactions, esterification and long chain polymerization. It acts as a monomer in the preparation of polyglycolic acid and other biocompatible copolymers. Further, it is used as a preservative and flavoring agent in organic synthesis. It is also used in adhesives and plastics.

Solubility
Soluble in water, methanol, diethyl ether, acetone, acetic acid and ethyl acetate.

Notes
Hygroscopic. Incompatible with bases, oxidizing agents and reducing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 79-14-1
Molecular Formula C2H4O3
Molecular Weight (g/mol) 76.051
MDL Number MFCD00004312
InChI Key AEMRFAOFKBGASW-UHFFFAOYSA-N
Synonym glycolic acid, hydroxyacetic acid, glycollic acid, hydroxyethanoic acid, acetic acid, hydroxy, glycolate, caswell no. 470, alpha-hydroxyacetic acid, kyselina glykolova, kyselina glykolova czech
PubChem CID 757
ChEBI CHEBI:17497
IUPAC Name 2-hydroxyacetic acid
SMILES C(C(=O)O)O

Specifications

pH 2
Melting Point 74°C to 80°C
Assay Percent Range 98%
Linear Formula HOCH2CO2H
UN Number UN3261
Beilstein 1209322
Merck Index 14,4498
Solubility Information Soluble in water,methanol,diethyl ether,acetone,acetic acid and ethyl acetate.
Formula Weight 76.05
Percent Purity 98%
Density 1.49
Quantity 100 g
Odor Unpleasant
Sensitivity Hygroscopic
Chemical Name or Material Glycolic acid
Show More Show Less
compliance-icons compliance-icons
Product Identifier
  • Glycolic acid
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 1
  • Skin corrosion/irritation Category 1 B
Hazard Statement
  • H314-Causes severe skin burns and eye damage.
  • H318-Causes serious eye damage.
  • H332-Harmful if inhaled.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.
Supplemental information
  • MIXTURE LIST-Contain: Hydroxyacetic acid

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.