missing translation for 'onlineSavingsMsg'
Learn More

Glycolic Acid, TRC

Product Code. 30136249
Change view
Click to view available options
Quantity:
1 g
10 g
100 g
Unit Size:
100g
10g
1g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30136249 10 g 10g
30134325 100 g 100g
17942935 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30136249 Supplier Toronto Research Chemicals Supplier No. TRCG64560010G
 Restricted Item

This item requires a price update prior to being ordered.

Request a Price for this item here.

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 79-14-1
Synonym Acetic acid, 2-hydroxy- (ACI), 2-Hydroxyacetic acid (ACI), Acetic acid, hydroxy- (9CI), Glycolic acid (7CI, 8CI), 2-Hydroxyethanoic acid, Glycocide, GlyPure, GlyPure 70, GlyPure 99, Hydroxyacetic acid, Hydroxyethanoic acid, NSC 166, YST-GCS, α-Hydroxyacetic acid
SMILES OCC(=O)O

Specifications

Molecular Formula C2 H4 O3
InChI Formula InChI=1S/C2H4O3/c3-1-2(4)5/h3H,1H2,(H,4,5)
IUPAC Name 2-hydroxyacetic acid
Molecular Weight (g/mol) 76.05
Formula Weight 76.016
Percent Purity >95%
Quantity 10 g
Chemical Name or Material Glycolic acid
Grade GC
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.